Anti-NLRP1 Rabbit Polyclonal Antibody

Leverancier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBIN0018-60HEA 526 EUR
USBIN0018-60H
Anti-NLRP1 Rabbit Polyclonal Antibody
Antilichamen
Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: NLRP1 (NLR family, pyrin domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR