Anti-SKIV2L2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverancier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBIS1014-10K2-APEA 852 EUR
USBIS1014-10K2-AP
Anti-SKIV2L2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antilichamen
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: SKIV2L2 (superkiller viralicidic activity 2-like 2 (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR