Anti-SRI Rabbit Polyclonal Antibody
Leverancier: United States Biological
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
USBIS5363-01AEA 539 EUR
USBIS5363-01A
Anti-SRI Rabbit Polyclonal Antibody
Antilichamen
Recommended dilution: Western Blot: 0.5-2?g/ml; Immunochemistry: 10 ?g/ml
Type: Primary
Antigen: SRI (sorcin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Horse, Human, Monkey, Mouse