Anti-T7 Chicken Polyclonal Antibody

Leverancier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBIT0230-02EA 434 EUR
USBIT0230-02
Anti-T7 Chicken Polyclonal Antibody
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Tag
Antigen: T7
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR