Anti-TCHHL1 Goat Polyclonal Antibody
Leverancier: United States Biological
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
USBIT8349-50AEA 551 EUR
USBIT8349-50A
Anti-TCHHL1 Goat Polyclonal Antibody
Antilichamen
Recommended dilution: ELISA: 1:4000; Western Blot: 1-3?g/ml
Type: Primary
Antigen: TCHHL1 (trichohyalin-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human