Anti-SCGB3A1 Goat Polyclonal Antibody

Leverancier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBIU7001-20AEA 420 EUR
USBIU7001-20A
Anti-SCGB3A1 Goat Polyclonal Antibody
Antilichamen
Recommended dilution: ELISA: 1:32000

Type: Primary
Antigen: SCGB3A1 (secretoglobin, family 3A, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR