Benchtop controller
Leverancier: Winkler
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WINKWRT2010X-WW010EA 740 EUR
WINKWRT2010X-WW010
Benchtop controller
Made of impact resistant polystyrene and used for equipment with multipole coupling and plug. It displays both the actual and setpoint digital temperatures.
  • Terminal block for signal contacts
  • Terminal block for sensors and analogue output
  • Fire protection class UL 94 HB
  • Integrated start and stop button
  • With built-in thermal fuse
Ambient temperature: 0 to 70 °C
Certificaten: DIN 53 453 and fire-protection class UL 94 H.