Netherlands
Order Entry
VWR®, Heating Blocks, Aluminium, Designed for 1,5 ml Tubes
Supplier:  VWR Collection
Certificates
undefined
VWR®, Heating Blocks, Aluminium, Designed for 1,5 ml Tubes
Product Information
  •  
    For
    No. of Holes
    Pk
  • Aluminium block
    Aluminium block
    460-0251
    0,5 ml tubes
    54
    1
    1 * 1 ST
    1 * 1 ST - 260,00 €
    260,00 €
    1 * 1 ST
       260,00 €
     : VWRI460-0251
    Specifications
    Environmentally Preferable
  • Aluminium block
    Aluminium block
    460-0250
    1,5 ml tubes
    40
    1
    1 * 1 ST
    1 * 1 ST - 260,00 €
    260,00 €
    1 * 1 ST
       260,00 €
     : VWRI460-0250
    Specifications
    Environmentally Preferable
  • Aluminium block
    Aluminium block
    460-0255
    0,5 and 1,5 ml tubes
    26 + 24
    1
    1 * 1 ST
    1 * 1 ST - 305,00 €
    305,00 €
    1 * 1 ST
       305,00 €
     : VWRI460-0255
    Specifications
    Environmentally Preferable
  • Aluminium block
    Aluminium block
    460-0256
    2,0 ml tubes
    40
    1
    1 * 1 ST
    1 * 1 ST - 260,00 €
    260,00 €
    1 * 1 ST
       260,00 €
     : VWRI460-0256
    Specifications
    Environmentally Preferable
  • Aluminium block
    Aluminium block
    460-0253
    15 mm tubes
    24
    1
    1 * 1 ST
    1 * 1 ST - 260,00 €
    260,00 €
    1 * 1 ST
       260,00 €
     : VWRI460-0253
    Specifications
    Environmentally Preferable
  • Aluminium block
    Aluminium block
    460-0252
    0,2 ml PCR tubes or plates
    96
    1
    1 * 1 ST
    1 * 1 ST - 260,00 €
    260,00 €
    1 * 1 ST
       260,00 €
     : VWRI460-0252
    Specifications
    Environmentally Preferable
  • Aluminium block
    Aluminium block
    460-0254
    Water bath block (115×73×38 mm)
    -
    1
    1 * 1 ST
    1 * 1 ST - 260,00 €
    260,00 €
    1 * 1 ST
       260,00 €
     : VWRI460-0254
    Specifications
    Environmentally Preferable
  • Aluminium block
    Aluminium block
    460-0257
    96-well ELISA plate
    -
    1
    1 * 1 ST
    1 * 1 ST - 305,00 €
    305,00 €
    1 * 1 ST
       305,00 €
     : VWRI460-0257
    Specifications
    Environmentally Preferable