Mesje voor veiligheidsmessen, nr. 45
Leverancier: Martor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
233-2211EA 9.8 EUR
233-2211
Mesje voor veiligheidsmessen, nr. 45
Scheermesjes
Industrial blade for use with safety knives and cutters.
  • Two-sided ground-edge
  • Single bevel edge-ground
  • Four-times usable blade
Mountable in:
ARGENTAX CLEVERCUT
SECUMAX EASYSAFE
SECUPRO OPTISAFE
SECUNORM HANDY
SECUNORM HANDYCUT
SECUNORM 175