Laminar flow workstations
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MSGA21740EA 0 EUR
MSGA21740 MSGA21741 MSGA21731 MSGA21742 MSGA21737 MSGA21738 MSGA21739 MSGA21733 MSGA21735
Laminar flow workstations
These laminar flow products provides cleanroom workplace with particle reduced air in a normaly ventilated area. Perfect for all areas where a particle free environment is required during processing, preparation or storage.
  • Modular construction with space-saving set up directly at the wall
  • Easy opening and closing of the sliding door
  • Corrosion-resistant, plastic laminated construction
  • Adjustable flow rate with flow indication
  • Multiple configuration with connections for central building control systems