Eyewash station, QuickSafe® Chemical Industry
Leverancier: Plum Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
QuickSafe®
118-0379EA 151 EUR
118-0379
Eyewash station, QuickSafe® Chemical Industry
Eerste-hulpbenodigdheden Oogdouches
This wall-mountable station is made of impact resistant plastic and equipped with a transparent cover. The station is specifically designed for use in the chemical industry, laboratories and in other companies where employees are in close contact with acids and alkalis.
  • Contains Duo eyewash to rinse both eyes at once
  • Cabinet is only slightly larger than an A3 sheet of paper
  • Comes with instructions and fixtures for wall mounting
Contents:
1×500 ml Plum Eye Wash Duo
1×200 ml pH Neutral
1×500 ml pH Neutral Duo
5×QuickRinse eyewash ampoules (20 ml)
1×QuickFix Elastic plaster refill
W×D×H: 253×92×430 mm