U heeft gezocht op: Ontgassers
5  results were found
SearchResultCount:"5"
Sort Results
List view Easy View (new)
Rate These Search Results
Catalogus nummer: (RHEO9000-1549)
Leverancier: IDEX HEALTH&SCIENCE LLC(RHEODY
Omschrijving: Dissolved gases and bubbles in system liquids cause dispense volume anomalies in many instruments, negatively affecting both dispense precision and an...
UOM: 1 * 1 ST
Catalogus nummer: (HPLC0001-6484)
Leverancier: COLLECTION PREFIX HPLC
Omschrijving: The Systec® Stand-Alone Prep-Scale HPLC vacuum degassing systems is high-efficiency, in-line modules that remove dissolved gases from the mobile phase...
UOM: 1 * 1 ST
Leverancier: COLLECTION PREFIX HPLC
Omschrijving: On-Line degassing has been shown to be more effective at removing dissolved gas from mobile phases than sonication or helium sparging. Up to five solv...
Catalogus nummer: (REST25194)
Leverancier: Restek
Omschrijving: Mobile phase degasser used to remove the dissolved gas from mobile phases. This is better than the sonication or helium sparging techniques.
UOM: 1 * 1 ST
Catalogus nummer: (REST25014)
Leverancier: Restek
Omschrijving: Bubbles in an LC system can cause check valve malfunctions and pump cavitation, seriously affecting pump performance. The debubbler removes bubbles fr...
UOM: 1 * 1 ST
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 5 of 5
no targeter for Bottom