U heeft gezocht op: Regelventielen
8  results were found
SearchResultCount:"8"
Sort Results
List view Easy View (new)
Rate These Search Results
Leverancier: IDEX HEALTH&SCIENCE LLC(RHEODY
Omschrijving: Rheodyne®, 7030L 2-position, 6-port switching valve, large bore, double 3-way, 10-32 ports for 1/16" OD tubing, 1,0 mm stator passage Ø, Roestvrij staal
Leverancier: IDEX HEALTH&SCIENCE LLC(RHEODY
Omschrijving: These high pressure actuated valves are designed to be added to an existing instrument or to work optimally with stand-alone lab configurations.
Catalogus nummer: (554-3288)
Leverancier: IDEX HEALTH&SCIENCE LLC(RHEODY
Omschrijving: Combine MX Series IIâ„¢ modular valves with an instrument currently in use to support complex fluid switching and sample injection needs.
UOM: 1 * 1 ST
Leverancier: IDEX HEALTH&SCIENCE LLC(RHEODY
Omschrijving: Applicable for sample injection, column switching and sample enrichment applications.
Leverancier: Cytiva
Omschrijving: Range of manual and automated valves to meet a variety of performance requirements. They are designed for biocompatibility and years of reliable servi...
Leverancier: UPCHURCH SCIENTIFIC
Omschrijving: Upchurch Scientific®, V-101L-60 right angle flow-switching valve, bulkhead version, 1/16" OD tubing, 1,0 mm thru-hole, 6,2 µl swept volume
Leverancier: OMNIFIT LTD
Omschrijving: The high pressure manual rotary valves are made from PTFE with a PCTFE rotor with a bore size of 0,8 mm making them ideal for pressure applications up...
Leverancier: OMNIFIT LTD
Omschrijving: Valves are constructed from PTFE with a PCTFE rotor and have a 1,5 mm bore size. They accept tubing sizes between 0,5 and 4 mm Ext.Ø. and are pressure...
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
1 - 8 of 8
no targeter for Bottom