Gas leak detector
Leverancier: Testo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TEST0632.3162EA 321 EUR
TEST0632.3162
Gas leak detector
This gas leak detector is designed for fast checking measurements.
  • Trend display shows maximum leakage
  • Optical presentation of the gas concentrations
  • Flexible measurement probe for inaccessible places
  • Earphone connection for secure leakage localisation in loud surroundings
Leveringsinformatie: Supplied with measurement probe, rechargeable battery and earphones.