Extension cables for temperature probes, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BOLA
BOHLP1724-38EA 179 EUR
BOHLP1724-38 BOHLP1724-16 BOHLP1724-46 BOHLP1724-08
Extension cables for temperature probes, BOLA
Kabels
PTFE, 4-pole measuring cable and Lemo coupling. It is suitable for all BOLA temperature probes with coupling type Lemo, socket size 1.
  • Excellent chemical resistance
  • Temperature resistant from -200 to +250 °C