Nitrile gloves, Venitactyl
- Catalog No:
- DELPV1420B10009
- DELPV1400B100/6.5
- DELPV1400B100/8.5
- DELPV1400B100/7.5
- DELPV1400PB100/7.5
- DELPV1400B100/9.5
- DELPV1420B1000/6.5
- Model:
- Venitactyl
- Venitactyl
- Venitactyl
- Venitactyl
- Venitactyl
- Venitactyl
- Venitactyl
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powdered
- Powder-free
- Powder-free
- Powder-free
- Powdered
- Powder-free
- Powdered
- EN norm:
- EN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS
- EN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS
- EN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS
- EN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS
- EN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS
- EN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS
- EN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS
- Food contact approved:
- +
- +
- +
- +
- +
- +
- +
- ModelGlove sizeColourLengthPackedPk
- DELPV1420B1000/6.5V1420B1000/6.5Venitactyl6,5Blue240 mm100/box1001 * 100 ST11,80 €
Specifications
ModelVenitactylAmbidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowderedEN normEN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUSFood contact approved+ - DELPV1400PB100/7.5V1400PB100/7.5Venitactyl7,5Blue240 mm100/box1001 * 100 ST21,20 €
Specifications
ModelVenitactylAmbidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowderedEN normEN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUSFood contact approved+ - DELPV1420B10009V1420B10009Venitactyl9,5Blue240 mm100/box1001 * 100 ST11,80 €
Specifications
ModelVenitactylAmbidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowderedEN normEN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUSFood contact approved+ - DELPV1400B100/6.5V1400B100/6.5Venitactyl6,5Blue240 mm100/box1001 * 100 ST20,10 €
Specifications
ModelVenitactylAmbidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeEN normEN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUSFood contact approved+ - DELPV1400B100/7.5V1400B100/7.5Venitactyl7,5Blue240 mm100/box1001 * 100 ST20,10 €
Specifications
ModelVenitactylAmbidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeEN normEN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUSFood contact approved+ - DELPV1400B100/8.5V1400B100/8.5Venitactyl8,5Blue240 mm100/box1001 * 100 ST20,10 €
Specifications
ModelVenitactylAmbidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeEN normEN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUSFood contact approved+ - DELPV1400B100/9.5V1400B100/9.5Venitactyl9,5Blue240 mm100/box1001 * 100 ST20,10 €
Specifications
ModelVenitactylAmbidextrous / hand-specificAmbidextrousEnvironmentally PreferableDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeEN normEN 455-1/2-/3, EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUSFood contact approved+