Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverancier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CAYM10004302-1EA 124 EUR
CAYM10004302-1
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antilichamen
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish PE (Phycoerythrin)roxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR