Bags, stand-up, PAPER/ALU/PE, Doypack®
Leverancier: Flexico
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MINIITM080303_U
Bags, stand-up, PAPER/ALU/PE, Doypack®
Bags, zip seal, food contact.
  • Moisture and oxygen barrier
  • Ideal for dry, humid and aqueous product
  • Suitable for food contact
  • Zip seal with sound and tactile feel
  • Resealable and reusable
  • Upright position for shelf display
The multi-layer PAPER/ALU/PE film provides excellent preservation of the content. The kraft paper gives the bag an authentic and natural look. The aluminum layer ensures preservation of your product freshness and gives it the best protection against humidity, UV and oxygen. It also guarantee flavours conservation.
Certificaten: Suitable for food contact. Certificate available on request.