PCR clean up kit, Cyclo-Prep™
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
K600-50RXNEA 272 EUR
VWRCK600-50RXN
PCR clean up kit, Cyclo-Prep™
Cyclo-Prep PCR Clean-Up Kit is a spin column-based purification kit for the rapid isolation of PCR fragments following amplification.
  • Rapid isolation of PCR fragments
  • Fast spin column format
  • Pure product ideal for cloning and sequencing
After clean up, ultra-pure isolated DNA may be used for restriction enzyme digestion, labelling, hybridisation, ligation, DNA cloning and fluorescent sequencing protocols. The Cyclo-Prep™ PCR Clean Up Kit contains all necessary reagents for DNA purification with no hazardous phenol or chloroform, making it both safe and effective.
Leveringsinformatie: Includes binding buffer, 55 ml; wash buffer, 16 ml; elution buffer, 10 ml; spin columns, (50); and collection tubes (50). Contains material for 50 isolations.