Standard mantory signs
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
116-0527EA 7.25 EUR
116-0527 BRDY818280 116-0526 116-0529 116-0528 BRDY819016 BRDY819019 116-0530 116-0531 116-0534 116-0536 116-0535 116-0516 116-0515 116-0518 116-0517 116-0519 BRDY818335 BRDY818334 BRDY818336 BRDY818339 BRDY818415 116-0521 116-0520 116-0523 116-0522 116-0525 116-0524 116-0509 116-0508 BRDY818420 BRDY818423 BRDY819314 BRDY818865 BRDY818867 BRDY818418 BRDY818417 BRDY818419 116-0510 116-0512 116-0511 116-0514 116-0513 116-0537 BRDY818271 BRDY819165 BRDY819761 BRDY818430 BRDY818276 BRDY818278 BRDY818277 BRDY818279 BRDY818117 BRDY818116 BRDY818429 116-0540 116-0543 116-0542
Standard mantory signs
Pictogrammen
Mantory signs have always a blue circular backround which contains a white pictogram.These signs display a message or action that must be carried out in a designated area.
  • Laminated polyester
  • Resistance to most chemical products and solvents
  • Resistance against extreme temperatures
  • Good weatherability and UV resistance
  • Excellent adhesion to low surface energy surfaces
  • Thickness: 0,1 mm
  • Different sizes