Portable gas leak detector
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PERCN9306089EA 1430 EUR
PERCN9306089
Portable gas leak detector
Lekdetectiesystemen Gaslekdetectiesystemen
Designed for use with gas chromatography (GC) systems, it detects minute leaks of any gas with a thermal conductivity different from air.
  • Rugged side grips for added durability
  • Automatic shut-off capabilities
  • Optimised sample flow path
  • Leak indicated by both an LED light display and an audible alarm
The reference gas inlet draws in ambient air for comparison to air drawn into the sample probe.
Detectable gases: Helium, nitrogen, argon, carbon dioxide, hydrogen.