Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
Heat resistant gloves, Terrytop
Leverancier: Honeywell Safety Products
Heat resistant terry cotton knitted gloves that provide protection against temperatures between 100 and 250 °C. The knitting technique helps to produce gloves that conform snugly to the hand, with good dexterity. Cotton provides mechanical resistance and insulation with respect to contact heat.
  • Maximum comfort and added sturdiness are ensured
  • 5-finger gloves for a better grip of hot objects as well as added dexterity
  • Cuff helps to protect the forearm from mechanical and thermal hazards
Terry
Knitted cotton gloves with ribbed cotton wrist. Ideal for handling warm parts in glass industries. Suitable for contact temperatures up to 150 °C.
EN 388 (1232), EN 407 (X2XXXX)
Terry Heavy
Knitted cotton gloves with winged thumb and long cotton cuff. Ideal for handling in glassworks, metallurgy and foundries. Suitable for mechanical and thermal hazards up to 250 °C.
EN 388 (2331), EN 407 (X2XXXX), Cat. II
Certificaten: EN 388 (1232), EN 407 (X2XXXX); Cat. II
Terry Lined
Knitted cotton gloves with winged thumb, long cuff and Jersey lining. Ideal for handling in glassworks, metallurgy and foundries. Suitable for mechanical and thermal hazards up to 250 °C.
EN 388 (1221), EN 407 (X2XXXX), Cat. II
Terry Mix
Cotton/polyester blend terry knitted gloves with elasticated, knitted wrist. Ideal for handling sharp and hot parts in dry environments. Suitable for contact temperatures up to 150 °C.
EN 388 (233X), EN 407 (X2XXXX)
login om uw prijzen en beschikbaarheid te zien
  Onthoud mij op dit apparaat
Order
Expand All / Collapse All
VPE VWR catalogus nummer Hoeveelheid
1 PAIR DALLRGT1685_U Unavailable
unavailable Dit artikel is vervallen en leverbaar zolang de voorraad strekt. Binnen de VWR catalogus kunt u zoeken op alternatieven. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Specificaties
Ambidexter/handspecifiek Hand-specific
Manchetstijl Knit wrist
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid