Single test cassettes for urine screening
Leverancier: SureScreen Diagnostics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720-1272EA 0 EUR
720-1272 700-9563 SUREMAMPC300 720-1265 720-0742 720-0654 SUREAMC300 720-1303 720-0745 720-0767 720-0734 SUREPPXC SURETRAMC 720-0758 720-0727 700-9543 700-9568
Single test cassettes for urine screening
Drugstests
Single test cassettes are an effective way to screen for specific drugs in urine when a small urine sample is received. With high accuracy, ease of use and results that you can trust every time, results are given in just 3 minutes. Each individual cassette is provided with a pipette and packaged in a foil pouch, to ensure the high quality of the product is retained throughout the shelf-life.
  • Ideal for detecting recent drug use
  • More than 20 different drug tests available
  • Easy to perform test routine
  • Clear results in 3 minutes
  • Greater than 99% accuracy
  • Pipette provided for easy sampling
Recommended markets and use:
On-site screening
Safety critical situations
Random screening
For-cause screening
Monitoring rehabilitation
Drugs to test for can include:
AMP - Amphetamines
BAR - Barbiturates
BUP - Buprenorphine
BZO - Benzodiazepines
COC - Cocaine
EDDP - Methadone Metabolite
FEN - Fentanyl
KET - Ketamine
MAMP - Methamphetamines
MDMA - Ecstasy
MOP - Morphine
MTD - Methadone
OPI - Opiates
OXY - Oxycodone
PCP - Phencyclidine
PPX - Propoxyphene
TCA - Tricyclic Antidepressants
TRA - Tramadol
THC - Cannabis