Ovulation test
Leverancier: SureScreen Diagnostics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SURELHS_U
Ovulation test
These test strips provide a quick and simple solution when testing for ovulation.
  • Easy to perform routine test with clear results in three minutes
  • Greater than 99% accuracy with 40 mIu/ml cut-off
  • High-quality of the product retained throughout the shelf-life
Verpakking: Individual strip in a foil pouch.
Opgelet: Should not be used as a contraceptive method.