Alcohol breath tube
Leverancier: SureScreen Diagnostics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SUREBS1/3EA 14 EUR
SUREBS1/3 SUREBS2 SUREBS1/20
Alcohol breath tube
Drugstests
The alcohol breath tube is a quick and cost-effective method for detecting alcohol in breath. Each test is individually wrapped, and detects the presence of alcohol at a concentration of 0,02% BAC. The result is read by observing a colour change.
  • Easy to use
  • Discreet size
  • Three different cut-off levels available (0,02%, 0,05% and 0,08%)
Recommended markets and use:
On-site screening
Safety critical situations
Random screening
For-cause screening
Monitoring rehabilitation