Pregnancy test
Leverancier: SureScreen Diagnostics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SUREGHCGSEA 61.9 EUR
SUREGHCGS
Pregnancy test
Diagnostische systemen Clinical Reagents and Kits
Human chorionic gonadotropin, or hCG, is a hormone produced by the placenta shortly after the implantation of a fertilized egg into the womb. The early appearance of hCG in both the urine and serum of pregnant women and its rapid rise in concentration during gestational growth make it an excellent marker for the early detection of pregnancy.
  • Detect hCG one day after the first missed menstrual period
  • Sensitive detection as low as 25 mIU/mL hCG in urine and/or serum specimens
  • Simple procedure: No reagent or equipment required
An extremely accurate pregnancy test for early diagnosis in a strip format.