Gas purification systems, Aeronex® I series
Leverancier: Entegris
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ENTEEGPS8IEA 0 EUR
ENTEEGPS8I ENTEEGPS4IAT ENTEEGPS4IA ENTEEGPS8IAT ENTEEGPS4I ENTEEGPS12I ENTEEGPS8IT ENTEEGPS4IT ENTEEGPS8IA
Gas purification systems, Aeronex® I series
These purifiers are designed for use with N₂ gas. They remove O₂, CO, CO₂, H₂O and non-methane hydrocarbons to sub-ppb levels in inert gases. These are used in Photolithography, MOCVD, ALD and LTE.
  • Complete automatic operation
  • System uses two purifier beds in order to maintain a continuous flow of pure gas
  • Removes contaminants to sub-ppb (part-per-billion) and ppt (part-per-trillion) levels
  • Improves system safety by locating all purifiers in one cabinet, with multiple safety controls
  • Power failure will not damage the purification system
  • Does not release hydrocarbons that may contaminate the process
  • Uses ambient temperature purification technology – no power or heat required
CE and SEMI® S2 certified