Frames for shelving systems, MetroMax i® and MetroMax Q®
Supplier: Metro International
Certificates
About this item
The corrosion resistant taupe epoxy finish frames with structural steel tubing and tri-lobal adapters to attach to posts are ideal for use when configuring work tables or as a 3-sided ledge to contain bulky items.
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- Frame, 3-sided, L×W: 610×610 mmSupplier Article Number: M3TF2424ELocal Article Number: METOM3TF2424ESpecifications:
Specifications
Environmentally Preferable - Frame, 3-sided, L×W: 1524×610 mmSupplier Article Number: M3TF2460ELocal Article Number: METOM3TF2460ESpecifications:
Specifications
Environmentally Preferable