Paint inspection gauge, Super Pig III
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIMESP1100EA 808 EUR
SIMESP1100
Paint inspection gauge, Super Pig III
Coatinglaag testers
This inspection gauge is a destructive precision tool for inspection and thickness measurement, on single or multiple coats, on virtually all substrates, including wood, plastics, metals.
  • Titanium anodised aluminum
  • Three knife sizes and one crosscut knife in one holder
  • Ergonomic design with revolving system
  • Bright white LED ensure clear vision through the microscope
  • Small size eases use in corners
  • Engraved scaling (D-factor) for easy calculations
Observes and measures substrate and film defects. It is a very stable instrument, also the integrated microscope has an excellent focus.
The microscope is provided with a double scale (mm and inch) which allows you to calculate to micrometers and mils.
Leveringsinformatie: Supplied with black marker, cutters (1, 2, 4 ), hexadiagonal wrench and black leather case with belt clip.