Straw pipettes, STRAWS 180
Leverancier: Grade
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GRADSTW180EA 137 EUR
GRADSTW180
Straw pipettes, STRAWS 180
Pipetten Rietpipetten
These disposable pipette straws are specifically designed for use with laboratory blender bags. They are sterile, unbreakable and eliminate the hazard of broken glass. They also avoid exposure to infectious materials and contaminating samples.
Length: 180 mm
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904