VWR® Bottles, Narrow Neck, with Thread
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
VWRLSTL17BEA 1410 EUR
VWRLSTL17B VWRLSTL10 VWRL080215 VWRL080216 VWRLSTL18B VWRLSTL12 VWRLSTL33 VWRLSTL22 VWRLSTL18 VWRLSTL12B VWRLSTL17 VWRLSTL10B
VWR® Bottles, Narrow Neck, with Thread
Flessen Glazen flessen
Glass, clear or amber.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • Fitted White PTFE lined cap and label where stated
Excellent for sampling, storing and transporting liquids. Amber coloured to protect light-sensitive materials.