Avantor® ACE® C18-Amide, HPLC/UHPLC Columns, 5 µm
Supplier: Avantor
Certificates
About this item
Avantor® ACE® C18-Amide is a uniquely designed stationary phase offering improved retention of polar compounds, enhanced phenolic resolution and alternative selectivity to C18 bonded columns. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
- Ultra-high purity, base deactivated silica with exceptional reproducibility
- Ideal for a wide range of analytes differing in hydrophobicity
- Wide range of particle sizes (2,0 to 10 µm) and complementary phases
- Low bleed for UV and LC/MS compatibility
- C18 hydrophobic plus Amide interactions, compatible with 100% aqueous conditions
- Method validation kits contain three different batches of the same phase - available on request
The special design of the ACE® C18-Amide ligand incorporates an extended alkyl chain spacer between the silica surface and amide polar group, which gives extra phase stability.
Specifications
- Catalog No:
- ACE-1212-1510
- ACE-1212-7520
- ACE-12120110GD
- ACE-1212-2520
- ACE-1212-2510
- ACE-1212-1520
- ACE-1212-1030
- ACE-1212-7530
- ACE-1212-0530
- ACE-1212-0103GD
- ACE-1212-1530
- ACE-1212-3030
- ACE-1212-2504
- ACE-1212-2530
- ACE12120120GD
- ACE-1212-0520
- ACE-1212-0510
- ACE-1212-1020
- ACE-1212-1010
- ACE-1212-1220
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 16,4%
- 16,4%
- 16,4%
- 16,4%
- 16,4%
- 16,4%
- 16,4%
- 16,4%
- 16,4%
- 16,4%
- 16,4%
- 16,4%
- 16,4%
- 16,4%
- 16,4%
- 16,4%
- 16,4%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Specifications
Related Information
Product Information
- Catalog #PhaseColumn FormatColumn hardwarePore sizeParticle SizeØ int.AvailabilityPrice
- Local Article Number: ACETACE-1212-1510Specifications:
Low Manufacturing Impact More
Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16,4%EndcappedYesMax. pressurepH range2 - 8Surface area300 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Local Article Number: ACETACE-1212-7520Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load16,4%EndcappedYesMax. pressurepH range2 - 8Surface area300 m²/gUSP codeL1Environmental certification - 1/16" connections1/16" connectionsACE-1212-1010C18-AmideSemi-preparativeStainless steel100 Å5 µm10,00 mmLog in for availability1 * 1 STLocal Article Number: ACETACE-1212-1010Specifications:
Low Manufacturing Impact More
Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16,4%EndcappedYesMax. pressure275 barpH range2 - 8Surface area300 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - 1/16" connections1/16" connectionsACE-1212-2510C18-AmideSemi-preparativeStainless steel100 Å5 µm10,00 mmLog in for availability1 * 1 STLocal Article Number: ACETACE-1212-2510Specifications:
Low Manufacturing Impact More
Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16,4%EndcappedYesMax. pressure275 barpH range2 - 8Surface area300 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - 1/16" connections1/16" connectionsACE-1212-1020C18-AmidePreparativeStainless steel100 Å5 µm21,20 mmLog in for availability1 * 1 ST3 140,00 €Local Article Number: ACETACE-1212-1020Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load16,4%EndcappedYesMax. pressure275 barpH range2 - 8Surface area300 m²/gUSP codeL1Environmental certification - 1/16" connections1/16" connectionsACE-1212-1520C18-AmidePreparativeStainless steel100 Å5 µm21,20 mmLog in for availability1 * 1 STLocal Article Number: ACETACE-1212-1520Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load16,4%EndcappedYesMax. pressure275 barpH range2 - 8Surface area300 m²/gUSP codeL1Environmental certification - 1/16" connections1/16" connectionsACE-1212-2520C18-AmidePreparativeStainless steel100 Å5 µm21,20 mmLog in for availability1 * 1 ST5 690,00 €Local Article Number: ACETACE-1212-2520Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load16,4%EndcappedYesMax. pressure275 barpH range2 - 8Surface area300 m²/gUSP codeL1Environmental certification - 1/16" connections1/16" connectionsACE-1212-0520C18-AmidePreparativeStainless steel100 Å5 µm21,20 mmLog in for availability1 * 1 STLocal Article Number: ACETACE-1212-0520Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load16,4%EndcappedYesMax. pressure275 barpH range2 - 8Surface area300 m²/gUSP codeL1Environmental certification - 1/16" connections1/16" connectionsACE-1212-1030C18-AmidePreparativeStainless steel100 Å5 µm30,00 mmLog in for availability1 * 1 ST6 840,00 €Local Article Number: ACETACE-1212-1030Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load16,4%EndcappedYesMax. pressure275 barpH range2 - 8Surface area300 m²/gUSP codeL1Environmental certification - Local Article Number: ACETACE-1212-7530Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load16,4%EndcappedYesMax. pressurepH range2 - 8Surface area300 m²/gUSP codeL1Environmental certification - Local Article Number: ACETACE-1212-0530Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load16,4%EndcappedYesMax. pressurepH range2 - 8Surface area300 m²/gUSP codeL1Environmental certification - 1/16" connections1/16" connectionsACE-1212-2504C18-AmideAnalyticalStainless steel100 Å5 µm4,00 mmLog in for availability1 * 1 STLocal Article Number: ACETACE-1212-2504Specifications:
Low Manufacturing Impact More
Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16,4%EndcappedYesMax. pressure275 barpH range2 - 8Surface area300 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Local Article Number: ACETACE-1212-1530Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load16,4%EndcappedYesMax. pressurepH range2 - 8Surface area300 m²/gUSP codeL1Environmental certification - Local Article Number: ACETACE-1212-3030Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load16,4%EndcappedYesMax. pressurepH range2 - 8Surface area300 m²/gUSP codeL1Environmental certification - Local Article Number: ACETACE-1212-2530Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load16,4%EndcappedYesMax. pressurepH range2 - 8Surface area300 m²/gUSP codeL1Environmental certification - Local Article Number: ACETACE-1212-0510Specifications:
Low Manufacturing Impact More
Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load16,4%EndcappedYesMax. pressurepH range2 - 8Surface area300 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Local Article Number: ACETACE-1212-1220Specifications:
More
Accessories
Specifications
Environmentally PreferableCarbon load16,4%EndcappedYesMax. pressurepH range2 - 8Surface area300 m²/gUSP codeL1Environmental certification
Guard cartridges, ACE HPLC guard holder may be required
Product Information
- Catalog #PhaseColumn FormatColumn hardwarePore sizeParticle SizeØ int.AvailabilityPrice
- For semi-preparative columns (I-Ø 7,75, 10,0, 21,2 mm)For semi-preparative columns (I-Ø 7,75, 10,0, 21,2 mm)ACE-12120110GDC18-AmideGuard cartridge100 Å5 µmLog in for availability1 * 3 ST656,00 €Local Article Number: ACETACE-12120110GDSpecifications:
Low Manufacturing Impact More
Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedMax. pressurepH rangeSurface areaUSP codeEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - For analytical columns (I-Ø 3,0, 4,0, 4,6 mm)For analytical columns (I-Ø 3,0, 4,0, 4,6 mm)ACE-1212-0103GDC18-AmideGuard cartridge100 Å5 µmLog in for availability1 * 5 ST255,00 €Local Article Number: ACETACE-12120103GDSpecifications:
Low Manufacturing Impact More
Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedMax. pressurepH rangeSurface areaUSP codeEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - For preparative columns (I-Ø 30,0 mm)For preparative columns (I-Ø 30,0 mm)ACE12120120GDC18-AmideGuard cartridge100 Å5 µmLog in for availability1 * 3 ST1 660,00 €Local Article Number: ACETACE12120120GDSpecifications:
Low Manufacturing Impact More
Accessories
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedMax. pressurepH rangeSurface areaUSP codeEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact
Guard cartridge holders and accessories
Product Information
- Catalog #DescriptionPkAvailabilityPrice
- Local Article Number: ACETC0001