Netherlands
Order Entry
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography Multi-Element standards, Supelco®
Supplier:  Merck
Certificates
undefined
Ion chromatography Multi-Element standards, Supelco®
Supplier:  Merck
Certificates
Specifications
  • M-Clarity quality level:
    MQ100
Specifications
Related Information
Product Information
  • Catalog #
    Supplier #
    Pk
    Availability
    Price
  • Ion chromatography anion multi-element standard solution I, (3 elements in water) 1000 mg/l: F, Br, PO4 Certipur®
    Ion chromatography anion multi-element standard solution I, (3 elements in water) 1000 mg/l: F, Br, PO4 Certipur®
    1.11437.0500
    1.11437.0500
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 99,20 €
    99,20 €
    1 * 500 mL
       99,20 €
    Supplier Article Number: 1.11437.0500
    Local Article Number: MERC1.11437.0500
  • Ion chromatography anion multi-element standard solution II, (3 elements in water) 1000 mg/l: Cl, NO3, SO4 Certipur®
    Ion chromatography anion multi-element standard solution II, (3 elements in water) 1000 mg/l: Cl, NO3, SO4 Certipur®
    1.11448.0500
    1.11448.0500
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 82,40 €
    82,40 €
    1 * 500 mL
       82,40 €
    Supplier Article Number: 1.11448.0500
    Local Article Number: MERC1.11448.0500
  • Ion chromatography IC anion multi-element standard solution I, (5 elements in water) 100 mg/l: F , 250 mg/l: Cl, 500 mg/l: NO3 / 500 mg/l: SO4 / 1000 mg/l: PO4 Certipur®
    Ion chromatography IC anion multi-element standard solution I, (5 elements in water) 100 mg/l: F , 250 mg/l: Cl, 500 mg/l: NO3 / 500 mg/l: SO4 / 1000 mg/l: PO4 Certipur®
    1.70398.0500
    1.70398.0500
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 168,00 €
    168,00 €
    1 * 500 mL
       168,00 €
    Supplier Article Number: 1.70398.0500
    Local Article Number: MERC1.70398.0500
  • Ion chromatography anion multi-element standard solution V, (6 elements in water) 10 mg/l: F, Br / 50 mg/l: NO₃, PO₄ / 100 mg/l: Cl / 200 mg/l: SO₄, Certipur®
    Ion chromatography anion multi-element standard solution V, (6 elements in water) 10 mg/l: F, Br / 50 mg/l: NO₃, PO₄ / 100 mg/l: Cl / 200 mg/l: SO₄, Certipur®
    1.09032.0100
    1.09032.0100
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 251,00 €
    251,00 €
    1 * 100 mL
       251,00 €
    Supplier Article Number: 1.09032.0100
    Local Article Number: MERC1.09032.0100
  • Ion chromatography cation multi-element standard solution I, (5 elements in diluted nitic acid) 10 mg/l: NH₄ / 50 mg/l: K / 100 mg/l: Na, Ca, Mg, Certipur®
    Ion chromatography cation multi-element standard solution I, (5 elements in diluted nitic acid) 10 mg/l: NH₄ / 50 mg/l: K / 100 mg/l: Na, Ca, Mg, Certipur®
    1.09036.0100
    1.09036.0100
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 251,00 €
    251,00 €
    1 * 100 mL
       251,00 €
    Supplier Article Number: 1.09036.0100
    Local Article Number: MERC1.09036.0100
  • Ion chromatography cation multi-element standard solution I, (9 elements in diluted nitic acid) 100 mg/l: Li, Na, K, NH4, Mn, Ca, Mg, Sr, Ba Certipur®
    Ion chromatography cation multi-element standard solution I, (9 elements in diluted nitic acid) 100 mg/l: Li, Na, K, NH4, Mn, Ca, Mg, Sr, Ba Certipur®
    1.10322.0100
    1.10322.0100
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 241,00 €
    241,00 €
    1 * 100 mL
       241,00 €
    Supplier Article Number: 1.10322.0100
    Local Article Number: MERC1.10322.0100