Netherlands
Order Entry
Avantor® ACE®, Guard Cartridge Holders
Avantor® ACE®, Guard Cartridge Holders
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Avantor® ACE®, Guard Cartridge Holders
Supplier:  Avantor
   Sustainable Options Available
Specifications
  • Catalog No:
  • H0001
  • BIO-GRD-0001
  • H0002
  • H0011
  • H0009
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
Specifications
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • 1 cm stand-alone cartridge holder for 1,0 to 4,6 mm ID columns
    1 cm stand-alone cartridge holder for 1,0 to 4,6 mm ID columns
    H0001
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 161,00 €
    161,00 €
    1 * 1 ST
       161,00 €
    Local Article Number: ACETH0001
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Semi-preparative cartridge holder for 7,75 to 21,2 mm ID columns
    Semi-preparative cartridge holder for 7,75 to 21,2 mm ID columns
    H0002
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 457,00 €
    457,00 €
    1 * 1 ST
       457,00 €
    Local Article Number: ACETH0002
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Preparative guard cartridge holder for 30 mm ID columns
    Preparative guard cartridge holder for 30 mm ID columns
    H0009
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 801,00 €
    801,00 €
    1 * 1 ST
       801,00 €
    Local Article Number: ACETH0009
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • UHPLC guard cartridge holder for 2,1 to 4,6 mm ID columns
    UHPLC guard cartridge holder for 2,1 to 4,6 mm ID columns
    H0011
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 222,00 €
    222,00 €
    1 * 1 ST
       222,00 €
    Local Article Number: ACETH0011
    Specifications:
      Low Manufacturing Impact
    More
    Accessories
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Guard cartridge holder for ACE® UltraCore columns
    Guard cartridge holder for ACE® UltraCore columns
    BIO-GRD-0001
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 569,00 €
    569,00 €
    1 * 1 ST
       569,00 €
    Local Article Number: ACEUBIO-GRD-0001
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
    Environmental certification