Safety drum vents
Leverancier: Justrite
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
119-0234EA 78.6 EUR
JUSU28206 JUSU08101 119-0234 JUSU08006
Safety drum vents
Ventielen Overdrukventielen
These safety drum vents provide automatic pressure relief for drums between 0,20 bar but less than 0,55 bar. Depending on the model, vacuum relief is either automatic (on horizontally installed vents) or manual (on vertically installed vents).
  • Vents can prevent escape of flammable vapour in closed position
  • Includes flame arrester for added safety