Drum keys
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.08803.0001EA 149 EUR
1.08803.0001
Drum keys
Moersleutels
Drum keys are used to open barrels and drums with 2 and ³/₄" screw caps.