N-Methyl-2-pyrrolidone (NMP), SupraSolv® for headspace gas chromatography
Supplier: Merck
Certificates
About this item
CAS Number:872-50-4
:
Formula:C₅H₉NO
MDL Number:MFCD00003193
:99.13 g/mol
Merck Index:13,06140
Boiling Pt:202 °C (1013 hPa)
Melting Pt:–24 °C
Density:1.033 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:91 °C
Storage Temperature:Ambient
EINECS:212-828-1
:GHS08
:GHS07
Related Information
Certificates
Search by Lot NumberMerck Pharma Segment Brochure Merck SupraSolv® High Purity Solvents for Gas Chromatography
Product Literature
msds_102497
SDS
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: 1.02497.0500Local Article Number: MERC1.02497.0500
- Supplier Article Number: 1.02497.1000Local Article Number: MERC1.02497.1000
- Supplier Article Number: 1.02497.2500Local Article Number: MERC1.02497.2500