Surface cleaner, floors, TASKI Jontec Forward
Leverancier: Diversey
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TASKI Jontec
DIVY7513111EA 70.8 EUR
DIVY7513111
Surface cleaner, floors, TASKI Jontec Forward
Reinigingsmiddelen Detergenten en oppervlaktereinigers
Highly effective, alkaline floor cleaner, designed for use in scrubber driers.
  • Slightly alkaline, low foaming
  • High emulsifying power
  • Manual or machine application