Netherlands
Order Entry
Palette knives, Steriware®
Palette knives, Steriware®
Supplier:  Sampling Systems
Certificates
undefined
Palette knives, Steriware®
Supplier:  Sampling Systems
Related Information
Product Information
  • Catalog #
    Supplier #
    Sterile
    Length
    Blade length
    Blade width
    Colour
    Pk
    Availability
    Price
  • SAMP8175H-150
    8175H-150
    -
    260 mm
    150 mm
    40 mm
    White
    100
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 297,00 €
    297,00 €
    1 * 100 ST
       297,00 €
    Supplier Article Number: 8175H-150
    Local Article Number: SAMP8175H-150
    Specifications:
    Specifications
    Environmentally Preferable
  • SAMP8175H-150S
    8175H-150S
    +
    260 mm
    150 mm
    40 mm
    White
    100
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 362,00 €
    362,00 €
    1 * 100 ST
       362,00 €
    Supplier Article Number: 8175H-150S
    Local Article Number: SAMP8175H-150S
    Specifications:
    Specifications
    Environmentally Preferable
  • SAMP8175H-200
    8175H-200
    -
    300 mm
    200 mm
    45 mm
    White
    100
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 304,00 €
    304,00 €
    1 * 100 ST
       304,00 €
    Supplier Article Number: 8175H-200
    Local Article Number: SAMP8175H-200
    Specifications:
    Specifications
    Environmentally Preferable
  • SAMP8175H-200S
    8175H-200S
    +
    300 mm
    200 mm
    45 mm
    White
    100
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 327,00 €
    327,00 €
    1 * 100 ST
       327,00 €
    Supplier Article Number: 8175H-200S
    Local Article Number: SAMP8175H-200S
    Specifications:
    Specifications
    Environmentally Preferable