VWR®, Antifoam emulsion (10% active) pH 5 - 7
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
632137GEA 0 EUR
VWRC632137G VWRC632136F VWRC632134D
VWR®, Antifoam emulsion (10% active) pH 5 - 7
Non ionic, water-based 10% active textile emulsion.
  • Immediate foam knock-down
  • Long-term foam inhibition
  • Low addition levels
  • Rapid dispersion in aqueous foaming systems
  • Stable in textile processes at pH 4 to 11
  • Foam control up to 100 °C
  • Good compatibility with dye stuffs, surfactants and textile auxiliaries