VWR®, Antifoam emulsion (100% active)
Leverancier: Avantor
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
632484WEA
93
EUR
632484W
VWR®, Antifoam emulsion (100% active)
Reinigingsmiddelen
Detergenten en oppervlaktereinigers