BOD bottles
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HACH62124EA 741 EUR
HACH62124 HACH62106 HACH62100
BOD bottles
Flessen BOD-flessen
These are standard bottles, made of borosilicate glass, for BOD analaysis.
  • Flared lip and ground-glass stopper to prevent air entrapment and for water seal
  • Serialised bottles have silk-screened, sequential numbers and include a bar code
  • Frosted marking area
Leveringsinformatie: Stoppers are included.