UHPLC columns, Hypersil GOLD™ Vanquish™ PFP
Supplier: Thermo Fisher Scientific
Certificates
About this item
These reversed phase columns were developed in conjunction with the Vanquish™ UHPLC system to take advantage of the system's extended pressure capabilities and robustness. The result is a new level of separation, speed and throughput that solve analytical challenges of analysing complex matrices using liquid chromatography and LC-MS.
- High efficiency enables separation of very complex mixtures
- High efficiency at high flow rates
Limited Time Sale
Select items below are currently on sale
Specifications
- Catalog No:
- 555-5611
- 555-5608
- Endcapped:
- Yes
- Yes
- Max. pressure:
- 1500 bar
- 1500 bar
- Surface area:
- 220 m²/g
- 220 m²/g
- Temperature limit:
- 60 °C
- 60 °C
Specifications
Product Information
- Catalog #Supplier #Column FormatPhasePore sizeParticle SizeØ int.LengthAvailabilityPrice
- 555-560825402-102130-VAnalyticalPFP175 Å1,9 µm2,10 mm100 mmLog in for availabilityLog in for availability1 * 1 ST1 * 1 ST - 1 096,50 €1 096,50 €
Sale Valid until 31/07/2025
Sale Valid until 31/07/20251 * 1 ST1 096,50 €Specifications:More
Specifications
Environmentally PreferableEndcappedYesMax. pressure1500 barSurface area220 m²/gTemperature limit60 °C - 555-561125402-152130-VAnalyticalPFP175 Å1,9 µm2,10 mm150 mmLog in for availabilityLog in for availability1 * 1 ST1 * 1 ST - 1 207,00 €1 207,00 €
Sale Valid until 31/07/2025
Sale Valid until 31/07/20251 * 1 ST1 207,00 €Specifications:More
Specifications
Environmentally PreferableEndcappedYesMax. pressure1500 barSurface area220 m²/gTemperature limit60 °C