Hand cleanser, Estesol® Premium PURE
Leverancier: SC Johnson
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Estesol® Premium PURE
115-2278EA 71.2 EUR
115-2278 115-2277 115-2279
Hand cleanser, Estesol® Premium PURE
Huidverzorging Zepen en huidreinigingsmiddelen
Soap-free, light duty hand cleanser to remove general dirt. For use in general industrial environments. Can be used on sensitive skin.
  • Contains skin protectant Eucornol®
  • Contains a mild blend of surfactants (sugar surfactant and sulfosuccinats) for fast and effective hand cleaning with a large proportion of moisturisers
  • Soap-free, petroleum solvent-free, silicone-free, perfume- and preservative-free
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904