Sanitising foams, Stoko Refresh™ Sanitizer Foam (2-C)
Leverancier: Deb Stoko
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EVON34989EA 73.7 EUR
EVON34989 EVON35209
Sanitising foams, Stoko Refresh™ Sanitizer Foam (2-C)
Antimicrobial active foam for use without water.
  • Effective against bacteria (incl. MRSA, VRE) and fungi
  • Bacterial reduction: 99.999 %
  • Contains replenishing conditioner
  • Alcohol based but kind to the skin
  • Fragrance-free
Certificaten: EN 1500, EN 13727, EN 1650, EN 1040, EN 1276