Sanitising gels, Stoko Refresh™ Sanitizer Gel (2-A)
Leverancier: Deb Stoko
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EVON34986EA 51.2 EUR
EVON34986
Sanitising gels, Stoko Refresh™ Sanitizer Gel (2-A)
Gel for optimal hand disinfection without water with simultaneous skin care by means of a moisturising complex. Effective against bacteria (including MRSA), fungi and enveloped viruses. For use without water.
  • Effective against bacteria(incl. MRSA), fungi
  • Bacterial reduction: 99.999 %
  • Tuberculocid in accordance with EN 14348
  • Virucidal against enveloped viruses, effective against A/H1N1 virus in line with EN 14476
  • Contains glycerine and panthenol
  • Alcohol based but kind to the skin
  • Fragrance-free
Certificaten: EN 1500, EN 12791, EN 13727, EN 1276, EN 1650, HACCP