Ceramic protective tubes, one end closed
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30052.DYEA 182 EUR
30052.DY 30044.DY 30052.DU 30052.EE 30044.EE
Ceramic protective tubes, one end closed
Slangen Transfer Tubing
Pure Al2O3 Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.