Multibore insulating tubes, ceramic
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
32558.ELEA 134.16 EUR
32558.EL 32560.DZ 32554.EW
Multibore insulating tubes, ceramic
Slangen Transfer Tubing
Pure Al₂O₃ Aluminium oxide, AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.