Tube for heating coils, ceramic
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
33205.DMEA 153.92 EUR
33205.DM 33205.DR
Tube for heating coils, ceramic
Slangen Transfer Tubing
Pure Al₂O₃ Aluminium oxide, AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.