Direct Methanol Fuel Cell (DMFC) screener membrane electrode assembly
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45378.KSEA 322 EUR
ALFA45378.KS
Direct Methanol Fuel Cell (DMFC) screener membrane electrode assembly
  • MEA-5 layer
  • Plus membrane
Physical form: Fuel cell component.
Active area: 100 cm²